Learn More
Thermo Scientific Chemicals Field's Stain A
Codice prodotto. p-7082202
Click to view available options
Quantità:
10g
50g
Dimensione della confezione:
10g
50g
Codice prodotto. 11448507
Marca: Thermo Scientific Chemicals A18577.18
Richiesta di formato massivo o personalizzato
Edge
Used to stain blood smears to identify malarial parasites
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Stain
Quantità 50g