Learn More
Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic Pipette
Click to view available options
Include:
Pipette Stand Only
Pipetter, Pipette Stand
Dimensione della confezione:
Pezzo
Descrizione
Combine Thermo Scientific™ Finnpipette™ Novus i Multichannel Electronic pipettes with our adjustable pipette stand for an economical semi-automated system for rapid development and application of Mass Spectrometric Immunoassay (MSIA) methods. Each Novus i comes with a custom MSIA menu for further simplification of pipette programming that is complementary to MSIA applications.
Features:
- Simplify MSIA methods using Finnpipette Novus i pipette on the pipette stand
- New software that allows 1-999 aspirate and dispense cycles with one press on the pipette trigger
- Continuous battery charging of pipette on the adjustable stand during sample processing
- Include forward, reverse, diluting, stepper pipetting, mixing, sequential stepping and more; Nine aspirate/dispense speeds
Compatible with:
Thermo Scientific MSIA D.A.R.T.'S™ (ordered separately)
Specifica
Specifica
| Descrizione | Finnpipette Novus i adjustable pipetter stand |
| Product Type | Pipette Stand |
| Include | Pipette Stand Only |
| Product Line | Finnpipette Novus i |