Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità (sistema metrico):
2 L
4 L
9 L
Tipo:
Rinse Bottle
Wash Bottle
Wash Bottle A
Wash Bottle B
Waste Bottle
Dimensione della confezione:
Pezzo
Descrizione
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
- Subsitute bottles of different sizes for added convenience
- Wash and waste bottles include liquid level sensors
- 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
Specifica
Specifica
| Include | Cap with tubing and liquid level sensors |
| Capacità (sistema imperiale) | 304.32 oz. |
| Capacità (sistema metrico) | 9 L |
| Forma | Rectangular |
| Tipo | Waste Bottle |
| Da utilizzare con (apparecchiatura) | Wellwash Versa Microplate Strip Washer |