Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Bottles
Codice prodotto. p-4659509 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità (sistema metrico):
2 L
4 L
9 L
Tipo:
Rinse Bottle
Wash Bottle
Wash Bottle A
Wash Bottle B
Waste Bottle
Dimensione della confezione:
Pezzo
Codice prodotto. 11708006
Marca: Thermo Scientific™ N10812
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
  • Subsitute bottles of different sizes for added convenience
  • Wash and waste bottles include liquid level sensors
  • 4L wash and 9L waste bottles available for automation applications
ELISA-based assays, cell washing
TRUSTED_SUSTAINABILITY
Specifica
Include Cap with tubing and liquid level sensors
Capacità (sistema imperiale) 135.25 oz.
Capacità (sistema metrico) 4 L
Forma Round
Tipo Wash Bottle B
Da utilizzare con (apparecchiatura) Wellwash Versa Microplate Strip Washer