Learn More
Fisherbrand™ Large Capacity Screw-Top Pathology Containers
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Capacità (sistema metrico):
2.27 L
Altezza (sistema imperiale):
7 in.
N. per custodia:
12/Cs.
Dimensione della confezione:
pezzi 12
Descrizione
Large-capacity pathology containers manufactured from high-quality, high-density polyethylene feature a leak-resistant screw cap closure with a gasket to help maintain sample integrityduring storage and transportation.
- Secure, leak-resistant screw cap closure with gasket
- Available in four sizes to hold from small to large specimens
- All containers complete with handle for easy carrying
- 5-gallon container features a convenient handle for easy closing and opening
Specifica
Specifica
| Materiale | HDPE |
| Capacità (sistema imperiale) | 0.6 gal. |
| Capacità (sistema metrico) | 2.27 L |
| Tipo di chiusura | Screw Cap with Gasket |
| Altezza (sistema imperiale) | 7 in. |
| N. per custodia | 12/Cs. |