Learn More
Fisherbrand™ Polypropylene Microtiter Tubes
Sfoglia Tutto Fisherbrand ProdottiDescrizione
Ideal for sample dilution and/or mixing prior to transfer into microtiter plates
Also suitable for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagents. Choose plain tubes or tubes with frosted writing area.
- Superior-grade polypropylene tubes and packing racks are autoclavable and can be frozen to -80°C
- Sterile tubes are certified RNase-, DNase-, ATP-, DNA- and pyrogen-free
- Frosted tubes feature writing area that wraps around tube collar
- Capacity- 1.2mL
Order Info
Packed in bulk, or in convenient 8 x 12 compact tray packs or robotic racks. Compact Tray PacksSnug, smooth- fitting cover protects samples and enables stacking; With paper grid for easy indentification of contentsRobotic RacksSpecifically designed with 96-well footprint for robotic workstations; One-way lid design with alphanumeric grid enables easy sample identification; Condensation rings inside cover minimize cross-contamination; Tubes fit in straight, upright position
Specifica
Specifica
| Certificazione/Conformità | RNase, DNase, ATP, DNA, pyrogen-free |
| Autoclavabile | Autoclavable |
| Capacità (sistema metrico) | 1.2 mL |
| Colore | Natural |
| Materiale | Polypropylene |
| Da utilizzare con (applicazione) | HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, sample freezer storage or transport of specimens and reagent |
| Tipo di etichetta | Frosted Writing Area |
| Imballaggio | Tray Pack |
| Sterilità | Sterile |
| Graduated | No |
| Vedi altri risultati |