Learn More
LevGo™ Inoculation Picks
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Altezza (sistema metrico):
32.5 mm
32.51 mm
63.5 mm
152.4 mm
Size:
Large
Mini
Standard
Dimensione della confezione:
pezzi 1600
pezzi 2000
pezzi 500
Descrizione
- Available in three lengths and two needle sizes to suit a wide range of application needs.
- Naturally biodegradable, made from 100% U.S.A grown white birch wood
- Manufactured in an FDA-registered factory and comply with FDA standards for food contact
- Available in semi or needle point styles for delicate operations
- Autoclavable
- Disposable
Specifica
Specifica
| Materiale | White Birch Wood |
| Size | Large |
| Product Type | Toothpick |
| Quantità | 500 Pk. |
| Autoclavabile | Not Autoclavable |
| Altezza (sistema metrico) | 152.4 mm |
| Altezza (sistema imperiale) | 6 in. |