Learn More
OHAUS™ Fixed Multi-Purpose Clamps
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Dimensioni (sistema metrico) impugnatura:
0 to 69 mm
0 to 77 mm
Jaw Type:
2-Prong Single Adjust
3-Prong Dual Adjust
Materiale:
Nickel-plated
Stainless Steel
Dimensione della confezione:
Pezzo
Descrizione
- Extension arms with a 360° rotation allow placement of apparatus at various distances from lab-frame without compromising experiment integrity
- Swivel and fixed position clamps hold apparatus near the lab-frame or stand, and have a built-in integral holder for attaching to lab-frames or apparatus
- Available as single or dual-adjust in nickel-plated zinc or stainless steel
- Non-slip vinyl sleeves, and fiberglass covers for temperatures above 100°C (included with all 2 and 3-prong clamps)
Specifica
Specifica
| Tipo | Fixed Clamp |
| Materiale | Nickel-plated |
| Include | Non-slip vinyl sleeves and fiberglass covers |
| Descrizione | Clamp, Multi Purpose, CLM-FIXED2SZM |
| Lunghezza (sistema imperiale) | 5.24 in. |
| Lunghezza (sistema metrico) | 133 mm |
| Size | Medium |
| Dimensioni (sistema metrico) impugnatura | 0 to 77 mm |
| Jaw Type | 2-Prong Single Adjust |