Learn More
Grant Instruments™ Flask Tray for Grant Shaking Water Baths
Click to view available options
Da utilizzare con (apparecchiatura):
LSB12
LSB18
Dimensione della confezione:
Pezzo
Beschreibung
Include threaded holes to accept flask clamps or holder for deep well plates (=2ml). Grant Instruments™ Flask Tray for Grant Shaking Water Baths are compatible with SR plate holders and racks or can be used alone to accommodate bags and miscellaneous vessels.
- Accommodates flasks from 25 to 1000mL
- For use with the followng Shaking Water Bath models:
- LSB12
- LSB18
- OLS26
- Compatible with SH plate holders or SR racks
- TF26 Test tube tray also compatible with SR racks
- Can be used alone to accommodate bags and miscellaneous vessels.
Spezifikation
Spezifikation
| Capacità (sistema metrico) | 25 to 1000 mL |
| Da utilizzare con (apparecchiatura) | LSB18 |
| Tipo | Flask Tray |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur