Learn More
Proteintech FlexAble Biotin Antibody Labeling Kit for Human IgG
Sfoglia Tutto Proteintech Prodotti
Click to view available options
Quantità:
10 Tests
100 Tests
50 Tests
Dimensione della confezione:
10 reazioni
200 reazioni
50 reazioni
Descrizione
FlexAble Biotin Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate Biotin in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifica
Specifica
| Quantità | 10 Tests |
| Formato | Liquid |