Learn More
Proteintech FlexAble CoraLite Plus 750 Antibody Labeling Kit for Human IgG
Sfoglia Tutto Proteintech Prodotti
Click to view available options
Quantità:
10 Tests
100 Tests
50 Tests
Dimensione della confezione:
10 reazioni
200 reazioni
50 reazioni
Descrizione
FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 750 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifica
Specifica
| Quantità | 50 Tests |
| Formato | Liquid |