missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLI1 Monoclonal Antibody (7.3)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA1196
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MA1-196 detects FLI-1 in human and mouse samples. MA1-196 has been successfully used in western blot, immunofluorescence and ChIP assays.
ETS-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes ETS-1, ETS-2, ERG-1, ERG-2, ELK, E74, FLI-1, PU. 1 and PEA3. Members of the ETS gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localization signal sequence.
Specifica
| FLI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
| P26323, Q01543 | |
| Fli1 | |
| Recombinant human DHFR-FLICTER (C-terminal region of FLI-1) protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG3 |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| 7.3 | |
| Unconjugated | |
| Fli1 | |
| Ewing sarcoma breakpoint region 2; EWSR2; Fli1; Fli-1; Fli-1 proto-oncogene, ETS transcription factor; Friend leukemia integration 1; Friend leukemia integration 1 transcription factor; Friend leukemia virus integration 1; Proto-oncogene Fli-1; Retroviral integration site protein Fli-1; Sic1; SIC-1; transcription factor ERGB | |
| Mouse | |
| Protein A | |
| RUO | |
| 14247, 2313 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto