Learn More
Thermo Scientific™ Temperature Flow and Pressure Control Accessories
Codice prodotto. 12200519 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Tipo:
Flow Control Kit
Pressure Reducer
Da utilizzare con (apparecchiatura):
ThermoFlex 3500 - 5000
ThermoFlex 7500 - 24000
ThermoFlex 900 - 10000
Size:
1 in. NPT
1/2 in. NPT
3/4 in. NPT
Dimensione della confezione:
Pezzo
Codice prodotto. Tipo Da utilizzare con (apparecchiatura) Size unitSize
12200519 Pressure Reducer ThermoFlex 900 - 10000 1/2 in. NPT Pezzo
17198155 Pressure Reducer ThermoFlex 3500 - 5000 3/4 in. NPT Pezzo
15681765 Pressure Reducer ThermoFlex 7500 - 24000 1 in. NPT Pezzo
15560163 Flow Control Kit ThermoFlex 900 - 10000 1/2 in. NPT Pezzo
15510435 Flow Control Kit ThermoFlex 7500 - 24000 1 in. NPT Pezzo
5 options
Codice prodotto. 12200519
Fornitore: Thermo Scientific™ 342000000025
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Refine the pressure and flow characteristics of your Thermo Scientific™ recirculating chiller.
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
  • Designed to work with your Thermo Scientific recirculating chiller
  • Available in popular sizes that match your recirculating chiller connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller connections
  • Designed to match the operating range of your recirculating chiller
Additional questions? Please contact Thermo Scientific Technical support if you’re unsure how best to refine the flow and pressure of your recirculating chiller.
TRUSTED_SUSTAINABILITY
Spezifikation
Da utilizzare con (apparecchiatura) ThermoFlex 900 - 10000
Tipo di pompa Centrifugal Pump, Turbine Pump
Tipo Pressure Reducer
Size 1/2 in. NPT
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.