Learn More
Thermo Scientific™ Temperature Flow and Pressure Control Accessories
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Tipo:
Flow Control Kit
Pressure Reducer
Da utilizzare con (apparecchiatura):
ThermoFlex 3500 - 5000
ThermoFlex 7500 - 24000
ThermoFlex 900 - 10000
Size:
1 in. NPT
1/2 in. NPT
3/4 in. NPT
Dimensione della confezione:
Pezzo
Descrizione
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
- Designed to work with your Thermo Scientific recirculating chiller
- Available in popular sizes that match your recirculating chiller connections and interface with your process
- Take the guess work out of finding parts to match your recirculating chiller connections
- Designed to match the operating range of your recirculating chiller
Additional questions? Please contact Thermo Scientific Technical support if you’re unsure how best to refine the flow and pressure of your recirculating chiller.
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | ThermoFlex 7500 - 24000 |
| Tipo di pompa | Centrifugal Pump, Turbine Pump |
| Tipo | Flow Control Kit |
| Size | 1 in. NPT |