Learn More
Thermo Scientific™ Fluid Transfer Assembly, Pump Set
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Dimensioni dei tubi:
10 in. TPE 374, 19.7 in. Pharmed BPT
Tipi di connettore:
MPX
Dimensions:
1/2 in. ID x 3/4 in. OD
Dimensione della confezione:
pezzi 10
Descrizione
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifica
Specifica
| Dimensions | 1/2 in. ID x 3/4 in. OD |
| Stile dei tubi | 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) OD; 0.25 m (10 in.) TPE 374 to 0.5 m (19.7 in.) Pharmed BPT to 0.25 m (10 in.); TPE 374; MPX |
| Tipi di connettore | MPX |
| Product Type | Pump Set |
| Dimensioni dei tubi | 10 in. TPE 374, 19.7 in. Pharmed BPT |