Learn More
Cytiva Fluoro Rubber/NBR/Polyamide/Polyethylene/Polypropylene/Stainless Steel material AC Flow Adapter
Sfoglia Tutto Cytiva Prodotti
Click to view available options
Pressione:
14.5 psi
Dimensione della confezione:
Pezzo
Descrizione
The basic C columns are of fixed volume but can be upgraded by adding these flow adapters (which keep bed height constant and protect from insoluble particles) and a thermostatic jacket. Three adjustable flow adapters are available for the C column series (AC 10, AC 16, and AC 26), allowing the choice of bed height to suit the application. Flow adapters are available for each of the column diameters to allow optimal bed heights for each application, as are thermostatic jackets for precise temperature control.
Specifica
Specifica
| Temperatura (sistema metrico) max | 60°C |
| Quantità confezione | 1 Pack |
| Pressione | 14.5 psi |