Learn More
Uvex Fold-flat Mask silv-Air Premium 5220+ FFP2
Sfoglia Tutto Uvex Prodotti
Click to view available options
:
Filter Type: FFP2
Descrizione
General Features
- Individually packed
- Color: gray
- With exhalation valve
Safety Features
- Certified FFP2 face mask to EN 149:2001+A1:2009
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Additional activated carbon filter against odours, gases and vapours below threshold
- Cannot be reused
Comfort Features
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
- Adjustable head band offering easy handling and comfortable fit
- Flexible and adjustable nose clip
- Soft material edges for high wearing comfort
Specifica
Specifica
| Certifications/Compliance (Europe) | EN 149 (D marked) |
| Monouso | Single-use |
| Materiale | Polypropylene, Polyester |
| With Exhalation Valve | Sì |
| Efficienza filtro | FFP2 |
| Sterilità | Non-sterile |