Learn More
Cole-Parmer Food, Milk, and Dairy Tubing
Codice prodotto. 30053166 Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1 in.
1-1/2 in.
1-1/4 in.
1-3/4 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
2 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
7/16 in.
7/8 in.
Diametro (sistema imperiale) esterno:
1 in.
1-1/2 in.
1-1/4 in.
1-1/8 in.
1-3/8 in.
1/2 in.
1/4 in.
1/8 in.
11/16 in.
13/16 in.
15/16 in.
2 in.
2-1/2 in.
2-1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
Dimensione della confezione:
15m
Codice prodotto. 30053166
Marca: Cole-Parmer 5012170
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Smooth, nonporous bore inhibits particle entrapment
Crystal clear for visual inspection and flow monitoring. Non-toxic, odor-free, and taste-free. Smooth, non-porous and flexible tubing promotes a sanitary fluid path. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifica
Diametro (sistema imperiale) interno 5/8 in.
Diametro (sistema imperiale) esterno 13/16 in.
Metodo di sterilizzazione Yes
Colore Clear