Learn More
Poulten Graf™ Dispenser di sicurezza Fortuna™ Optifix™
Click to view available options
Volume (sistema metrico):
1 - 5 ml
10 - 50 ml
2 - 10 ml
Da 0.4 a 2 ml
Da 1 a 5 ml
Da 10 a 50 ml
Da 2 a 10 ml
Da 20 a 100 ml
Da 6 a 30 ml
Include:
Pistone integrato PTFE, coperchio sigillante
Pistone rivestito in PTFE, labbro di tenuta
Pistone rivestito in PTFE, rubinetto d'arresto e valvola di sfiato
Pistone speciale rivestito in robusto PTFE
Dimensione della confezione:
Pezzo
Descrizione
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
- Manufactured according to DIN EN ISO 9001:2008 using high quality materials
- With certificate, individually numbered
- Glass dispensing cylinder is coated with plastic to provide breakage protection
- Ergonomic finger protection cap made of PP with volume setting system
- Graduation on the scale is lasered and easy to read
- Fast and easy volume setting with additional fine adjustment
Specifica
Specifica
| Product Type | Erogatore di sicurezza |
| Volume (sistema metrico) | Da 0.4 a 2 ml |
| Include | Pistone rivestito in PTFE, labbro di tenuta |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu