missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10849-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The calculated molecular weight of FOXO3 is 71 kDa, but some experiments using a FOXO3a specific antibody showed the expression of a 90 kDa full-length FOXO3a protein (Full) and a 40 kDa processed FOXO3a product (Short).
FoxO (forkhead box O) proteins are a family of transcription factors characterized by the distinct forkhead domain. The FoxO proteins act in the insulin signaling pathway, but FoxO3 has also been shown to play a role in the regulation of apoptosis, cell cycle progression and cancer. Several splice variants of FoxO3 have been reported.Specifica
| FOXO3A | |
| Polyclonal | |
| Unconjugated | |
| FOXO3 | |
| AF6q21, AF6q21 protein, FKHRL1, FKHRL1P2, forkhead box O3, Forkhead box protein O3, FOXO2, FOXO3, FOXO3A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2309, 294515, 56484 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43524, Q9WVH4 | |
| Foxo3 | |
| FOXO3A Fusion Protein Ag1289 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto