missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-77175
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FOXO4 Polyclonal specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.
Specifica
| FOXO4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4303 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 10 μg/mL | |
| NP_005929 | |
| FOXO4 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human FOXO4. The immunogen is located within amino acids 50 - 100 of FOXO4. Amino Acid Squence: LPSRLPEPAGGPQPG | |
| 0.1 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto