missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Specifica
| Antigene | FOXO4 |
|---|---|
| Concentrazione | 0.23 mg/mL |
| Applicazioni | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
Descrizione
This gene encodes a member of the O class of winged helix/forkhead transcription factor family. Proteins encoded by this class are regulated by factors involved in growth and differentiation indicating they play a role in these processes. A translocation involving this gene on chromosome X and the homolog of the Drosophila trithorax gene, encoding a DNA binding protein, located on chromosome 11 is associated with leukemia. Multiple transcript variants encoding different isoforms have been found for this gene.Specifica
| FOXO4 | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P98177, Q9WVH3 | |
| 4303, 54601 | |
| Peptide | |
| Primary | |
| -20°C | |
| FOXO4 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AFX, AFX1, forkhead box O4, Forkhead box protein O4, FOXO4, MLLT7 | |
| Foxo4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto