Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 5 in/127 mm
Codice prodotto. 15753265
Codice prodotto. 15753265
Marca: Eppendorf™ 6001061510
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U101, CryoCube™ F101h, 5 in/127 mm, side access, stainless steel- Rack for 127 mm/5 in boxes (15/ 50 mL conical tubes)- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 290 x 142 x 274 (11.4 x 5.6 x 10.8 in)- Max. box number per rack: 4 (2 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 24 boxes)
TRUSTED_SUSTAINABILITY
Specifica
Product Type Freezer Rack