Learn More
Scilabub™ Frosters™ Cryogenic Shoulder Glove
Codice prodotto. p-8010090
Click to view available options
Size:
Small
Medium
Large
Dimensione della confezione:
1 paio
Codice prodotto. 12991185
Marca: Scilabub™ GLO/CMS/EX
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Frosters™ Polyimide Cryogenic Shoulder Gloves have been specially developed for the handling of very cold objects. Typically they are used for handling objects stored in the vapour phase of liquid nitrogen (ranging from -120°C to -190°C).
The outer layer is of waterproof material to resist condensed water on cold surfaces, but remains permeable to gases (including water vapour) so that there is no clammy condensation inside. To reduce bulk around the fingers, the stitch holes have not been sealed. In normal use this has not proved a problem, however, the gloves should not be immersed in liquid nitrogen and it is not advisable to use them above -80°C. Very durable, they retain their flexibility (albeit with some stiffening) for long periods in cold atmospheres. Polyimide outer material, polyolefin with some polyester as filling and cotton liner. Manufactured to EN511.Developed for handling very cold objects, typically those stored in the vapour phase of liquid nitrogen (in the range -120°C to -190°C).
Specifica
Certificazione/Conformità CE 0120, EN 5 11: 2006
Colore Blue
Product Type Cryogenic Frosters Mid-arm Gloves
Size Small
Resistente a Water