missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-77021
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FTO Polyclonal specifically detects FTO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| FTO | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79068 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9C0B1 | |
| FTO | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human FTO. The immunogen is located within the first 50 amino acids of FTO. Amino Acid Squence: TWLPYLTPKDDEFYQ | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto