Learn More
G418 disulfate, Cell Culture Reagent, Thermo Scientific™
G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic.
Marca:  Thermo Scientific Alfa Aesar J62671.14
Codice prodotto. 15454999
  • ONLINE EXCLUSIVE PRICE
    2079.00€
    2038.10€ / 25g
    Risparmia 40.90€
    1% Sconto
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General description
• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis
Applications
• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry
TRUSTED_SUSTAINABILITY
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS Documenti
Documenti
Certificati
Promozioni
Promozioni