Learn More
GABRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14104-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.Specifica
| GABRG2 | |
| Polyclonal | |
| Unconjugated | |
| GABRG2 | |
| CAE2, ECA2, GABA A receptor gamma 2, GABAA receptor gamma 2, GABRG2, GEFSP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14406, 2566, 29709 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P18507, P18508, P22723 | |
| Gabrg2 | |
| GABRG2 Fusion Protein Ag5237 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |