missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-4 Mouse anti-Human, Clone: 3C2G3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66686-1-IG-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GAL4 is a transcriptional activator which activates genes necessary for galactose catabolism. The DNA binding domain of GAL4 is contained in the first 74 amino acids and contains a "zinc finger" sequence motif. However, amino acids 1-147 have been shown to bind to DNA in vitro. While GAL4 (1-74) bind DNA specifically, it is with a lower affinity than the GAL4 (1-147). GAL4 (1-147) binds to a specific 17 base pair nucleotide sequence, but alone is insufficient to activate transcription. Either of the two acidic activating regions, activating region I (amino acids 148-238) or activating region II (amino acids 768-881), must be joined to activate transcription.Specifica
| Galectin-4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P56470 | |
| LGALS4 | |
| Galectin-4 Fusion Protein Ag4049 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 3C2G3 | |
| Unconjugated | |
| LGALS4 | |
| Antigen NY CO 27, Gal 4, GAL4, Galectin-4, L 36 lactose binding protein, L36LBP, Lactose binding lectin 4, LGALS4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3960 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction