Learn More
Tocris Bioscience™ Galiellalactone
Click to view available options
Quantità:
500 μg
Dimensione della confezione:
500µg
Descrizione
Galiellalactone is a STAT3 inhibitor. Irreversibly binds STAT3 and inhibits binding to DNA. Inhibits IL-6-dependent JAK/STAT signaling. Induces cell-cycle arrest in G2/M phase and apoptosis in androgen-insensitive prostate cancer cells via activation of ATM/ATR pathway. Also destabilizes microtubules and inhibits cell migration. Exhibits anticancer effects in a mouse tumor xenograft model.
Specifica
Specifica
| Product Type | Inhibitor |
| Proteine bersaglio | STAT |
| CAS | 133613-71-5 |
| Formula molecolare | C11H14O3 |
| Peso formulazione | 194.23g/mol |
| Content And Storage | Store at -20°C |
| Da utilizzare con (applicazione) | STAT Inhibitors |
| Quantità | 500 μg |
For Research Use Only