Learn More
WTW™ Sensore galvanico di ossigeno disciolto per misuratori di conduttività e multiparametro WTW™
Click to view available options
Lunghezza (sistema metrico) cavo:
1.5 m
10 m
15 m
3 m
6 m
Include:
1 exchange membrane head
3 exchange membrane heads
Soluzione per pulizia cavo L da 1.5 m, lamina abrasiva, 3 teste membrana intercambiabili ed elettrolita
Soluzione per pulizia cavo L da 3 m, lamina abrasiva, 3 teste membrana intercambiabili ed elettrolita
Dimensione della confezione:
Pezzo
Descrizione
- Dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring
- Can be used in the lab as well as in the field
- Features include a quick response time (t99<60 s) and low-maintenance operation
- Fields of use include surface waters, measurements in wastewater treatment plants, BOD directions
- Filled with electrolyte and achieves a long dwelling time up to 6 months
- Calibration vessel is part of the delivery
- For use with conductivity and multiparameter meters
- Includes Cable, Cleaning solution, abrasive foil, exchange membrane head(s) and electrolyte
Specifica
Specifica
| Lunghezza (sistema metrico) cavo | 15 m |
| Include | 3 exchange membrane heads |
| Da utilizzare con (apparecchiatura) | Conductivity and multiparameter meters |