Learn More
Ansell GAMMEX™ Guanto chirurgico in neoprene
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Dimensione della confezione:
200 paia
Descrizione
- Non-latex and 100% chemical accelerator-free
- Featuring SENSOPRENE™ Technology to deliver a thinner yet robust glove for added comfort and durability
- Up to 30% thinner on average than Ansell's standard neoprene surgical gloves
- Smooth with a micro-textured finish for secure instrument handling and grip
- Tested for use with chemotherapy drugs and a wide range of chemicals
Specifica
Specifica
| AQL | 0.65 |
| Colore | Natural |
| Monouso | Single-use |
| Finitura esterna | Fully Textured |
| Latex Free | Sì |
| Materiale | Neoprene |
| Powdered | Powder Free |
| Size | 5.5 |
| Sterilità | Sterile |
Titolo del prodotto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido