Learn More
GAPDH Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 52 publications
Marca: Novus Biologicals NB300-320
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat, Canine, Drosophila, Insect samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifica
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GAPDH | |
| Affinity Purified | |
| IgG |
| Polyclonal | |
| 36 kDa | |
| Store at -20C. Avoid freeze-thaw cycles. |