missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11711-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Required for ribosome biogenesis and telomere maintece. NOLA1 is part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequently attached to C5, instead of the normal N1. Each rRNA can contain up to 100 pseudouridine ("psi") residues, which may serve to stabilize the conformation of rRNAs. NOLA1 may also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.Specifica
| GAR1 | |
| Polyclonal | |
| Unconjugated | |
| GAR1 | |
| GAR1, NOLA1, snoRNP protein GAR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 499709, 54433, 68147 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6AYA1, Q9CY66, Q9NY12 | |
| GAR1 | |
| GAR1 Fusion Protein Ag2282 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto