missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATAD2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12294-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Methylation of cytosine at CpG dinucleotides is a common feature of many higher eukaryotic genomes. Increasing evidence indicates that recruitement of histone deacetylase complexes by methyl-CpG-binding proteins (MeCP) is a major mechanism of methylated DNA silencing. P66, a zinc finger-containing protein is one of the components of the MeCP1 complex. P66 and p68, the two components of MeCP1 are different forms of the same zinc finger-containing protein and are conserved from C.elegans to humans. Homologs of p66 from different organisms revealed two highly conserved regions, CR1 and CR2. While CR1 is involved in the association of p66 with other MeCP1 components, CR2 plays an important role in targeting p66 and MBD3 to specific loci.Specifica
| GATAD2A | |
| Polyclonal | |
| Unconjugated | |
| GATAD2A | |
| GATAD2A, Hp66alpha, p66 alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234366, 290669, 54815 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q86YP4, Q8CHY6 | |
| Gatad2a | |
| GATAD2A Fusion Protein Ag2947 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto