missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16317-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The degradation of L-threonine to glycine consists of a two-step biochemical pathway involving the enzymes L-threonine dehydrogenase and 2-amino-3-ketobutyrate coenzyme A ligase. L-Threonine is first converted into 2-amino-3-ketobutyrate by L-threonine dehydrogenase. GCAT is the second enzyme in this pathway, which then catalyzes the reaction between 2-amino-3-ketobutyrate and coenzyme A to form glycine and acetyl-CoA. The enzyme is considered a class II pyridoxal-phosphate-dependent aminotransferase.Specifica
| GCAT | |
| Polyclonal | |
| Unconjugated | |
| GCAT | |
| AKB ligase, Aminoacetone synthase, GCAT, Glycine acetyltransferase, KBL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23464, 26912, 366959 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75600, O88986 | |
| Gcat | |
| GCAT Fusion Protein Ag9367 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto