Learn More
Thermo Scientific™ GENESYS™ 140/150 Vis/UV-Vis Spectrophotometers
Codice prodotto. p-8430008 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Include:
Genesys 140/150 EU Power Cord
Genesys 150 + EU Power Cord
Larghezza di banda spettrale:
2 nm
5 nm
Wavelength Range:
190 to 1100 nm
325 to 1100 nm
Warranty:
2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Dimensione della confezione:
Pezzo
Codice prodotto. 15810002
Marca: Thermo Scientific™ 840308300
Get the most for your QA/QC or teaching lab with a rugged, network ready spectrophotometer designed for repetitive measurements and ease of use.
Perform quantitative measurements in routine industrial QA/QC or university chemistry labs with the automated and network-capable Thermo Scientific™ GENESYS™ 150 UV-Visible and GENESYS 140 Vis Spectrophotometers. Optimized for usability and performance, this platform features a high-resolution color touchscreen, optional Wi-Fi networking, and a rugged design built for repetitive, heavy use environments. Regarded worldwide for reliability, accuracy and reproducibility, GENESYS spectrophotometers meet today's expectations for advanced technology in a compact, robust package.
The GENESYS 150 UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience. A full line of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control.
The GENESYS 140 Vis Spectrophotometer is a visible spectral region version in the instrumental class. Both instruments feature data resolution from 0.2 nm to 5 nm.
Features:
  • 7-inch high-resolution touchscreen user interface
  • Onboard methods, reporting and data export
  • Networking capable (Wi-Fi ready)
  • Room light resistance for open-lid operation
  • Sipper accessory
  • Removable, washable sample compartment liner with magnetic placement and hold-down
  • Accommodates cells up to 100 mm pathlength
  • (Optional) 8-position cell changer with switchable turret
  • (Optional) 4-position cell changer (long path cells)
  • (Optional) Peltier-thermostatted single cell holder (20 °C - 60 °C)
  • (Optional) Accessory for test-tubes up to 25 mm diameter and 150 mm height
Designed for:
  • Advanced teaching labs
  • R&D departments
  • Applications requiring high throughput or temperature control
TRUSTED_SUSTAINABILITY
Specifica
Piattezza della linea di base ±0.005 A
Compartimenti 8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Collegamenti Single USB-A
Duplex USB-A
Export data to network or PC via USB, Ethernet, or Wi-Fi
Tipo di rilevatore Fotodiodi doppi al silicio
Display 7 in. High Definition Color Touchscreen, Tiltable, 800 x 1280 pixels
Dimensioni (L x P x A) 35.5 x 38.5 x 19.5 cm
Sorgente luminosa Tungsten Halogen
Larghezza di banda spettrale 5 nm
Wavelength Range 325 to 1100 nm
Deriva (At 500 nm after 1 hour warmup) <0.0005 A/hr.
Requisiti elettrici External AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100/240 V, 50/60 Hz
Include Genesys 140/150 EU Power Cord
Tastierino Touchscreen
Durata utile della lampada >1000 hrs.
Rumore (RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Design ottico Double Beam
Precisione fotometrica ±0.002 A at 0.5 A, ±0.004A at 1.0 A, ±0.008 A at 2.0 A
Display fotometrico -3A to +5A
Gamma fotometrica –3A to +3.5A
Ripetibilità fotometrica ±0.001 A at 1 A measured at 1.0 A at 546 nm
Stampante inclusa Snap-on printer available
Scan Speed Automatic – up to 1800 nm/min.
Luce dispersa <0.05%T at 340 nm and 400 nm
Warranty 2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Accuratezza della lunghezza d'onda ±0.5 nm
Intervallo dati lunghezza d'onda 0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Ripetibilità della lunghezza d'onda < ±0.2 nm
Peso (sistema metrico) 7.5 kg