Learn More
GIGYF2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24790-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The function of this protein remains unknown.Specifica
| GIGYF2 | |
| Polyclonal | |
| Unconjugated | |
| GIGYF2 | |
| GIGYF2, GYF2, KIAA0642, PARK11, PERQ2, PERQ3, TNRC15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26058 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6Y7W6 | |
| GIGYF2 | |
| GIGYF2 Fusion Protein Ag20329 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |