Learn More
DiscoverX GIP (Gastric Inhibitory Peptide)
Click to view available options
Quantità:
500 μg
Descrizione
Gastric inhibitory polypeptide (GIP) is a secretin family endocrine peptide hormone produced in the small intestine that induces insulin secretion in a glucose-dependent manner and in response to eating. It also weakly inhibits gastric acid secretion. Acts as an agonist for GIPR. Provided as 0.5mg of dry powder.
Specifica
Specifica
| Product Type | Control Ligands |
| Quantità | 500 μg |