Learn More
Hirschmann™ Matraccio volumetrico graduato in vetro marrone con stopper in polietilene
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
Marca: Hirschmann™ 2822165
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- With polyethylene stopper
- With amber stain graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifica
DIN EN ISO 1042 | |
20 mL | |
110 mm | |
10/19 NS Standard Ground | |
Sì |
Polyethylene | |
Vetro Duran, Duran Glass | |
±0.04 mL | |
±0.04 ml | |
A |