Learn More
Hirschmann™ Pipetta capillare in vetro
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
Marca: Hirschmann™ 9600150
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- With ringmark
- Conformity certified
- Manufactured from durable DURAN™ glass
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifica
0.0025 | |
Vetro | |
50 μl | |
Sì |
ISO 7550, CE, IVD 98/79 EC | |
Marchio a rullo | |
Pipetta capillare |