Learn More
Hirschmann™ Pipetta capillare in vetro
Excellent for working with small samples in the laboratory. Hirschmann Instruments™ ringcaps™ Disposable Capillary Pipettes can be utilized for a wide range of scientific and medical applications.
Marca: Hirschmann™ 9600199
Dettagli aggiuntivi : Peso : 0.48600kg
Descrizione
- With ringmark
- Conformity certified
- Manufactured from durable DURAN™ glass
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifica
0.25% | |
Glass | |
100 μL | |
Single-use |
ISO 7550, CE, IVD 98/79 EC | |
Ring Mark | |
Capillary Pipette |