Learn More
Brady™ Miniature Circuit Breaker Lockouts - Pin-Out Wide
A safe and effective method for locking out European Circuit Breakers.
Marca: Brady™ 090851
Dettagli aggiuntivi : Peso : 0.02540kg
Descrizione
- Requires no tools to install
- Lockout easily installs with the aid of a push button
- Tie-bar style lockout has a thumb wheel for quick application
Specifica
Circuit Breaker Lockout | |
Fiberglass-reinforced Plastic | |
Miniature Circuit Breaker Lockouts - Pin-Out Wide | |
6 Pk. |
Red/Black | |
UL 94 HB | |
Fiberglass-reinforced Plastic |