Learn More
Trajan Scientific and Medical™ Glass Lined Stainless-Steel GC Tubing
Codice prodotto. 15288993
Click to view available options
Overall Length (Metric):
30 cm
60 cm
90 cm
180 cm
Diametro (sistema metrico) interno:
0.3 mm
0.4 mm
0.5 mm
0.70 mm
0.75 mm
0.8 mm
1 mm
2 mm
4 mm
9.5 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15288993
Marca: Trajan Scientific and Medical™ 082737
Exhibits resistance to the harshest of conditions and to strong acids and bases
  • Ideal for HPLC columns, particularly for biological matrices
  • Temperature stable up to 500°C
Specifica
Da utilizzare con HPLC Columns, Biological Matrices
Tipo Tubing
Diametro (sistema metrico) interno 1 mm
Overall Length (Metric) 30 cm
Materiale Borosilicate Glass (Lining)
Temperatura operativa max (sistema metrico) Stable up to 500°C