Learn More
Fisherbrand™ Imbuto separatore a goccia in vetro con rubinetto d'arresto in vetro
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Capacità (sistema metrico):
50 ml
100 mL
250 ml
500 ml
1000 mL
Numero stopper:
19/26
24/29
29/32
Dimensione della confezione:
Pezzo
Descrizione
Separates mixtures into immiscible solvents. Fisherbrand™ Glass Pear-Shaped Separating Funnel with Glass Stopcock is available with interchangeable glass or PTFE stopcock, but supplied with a glass and plastic stopper.
- Easy-open stopper helps to reduce pressure buildup during processing.
- Constructed in chemical resistant borosilicate glass.
- ISO 4800 Certified.
Specifica
Specifica
| Capacità (sistema metrico) | 1000 mL |
| Materiale | Borosilicate Glass |
| Certificazione/Conformità | ISO 4800 |
| Quantità | 1PK |
| Valvola di chiusura | Glass |
| Stopper Material | Glass, Plastic |
| Numero stopper | 29/32 |
| Product Type | Separating funnel |