Learn More
Smeg™ Glassware GW1160SC Washer with Steam Condenser
Click to view available options
:
Glassware GW1160SC Washer with Steam Condenser
Descrizione
- Washing up to 95°C
- Microprocessor programmable electronic control
- Serial port RS232 for connection to external printer or PC
- High efficency steam condenser Ecoslim, with cold water injection
- Peristaltic pump for liquid detergent, acid liquid neutralizer
- Installing one additional peristaltic pump possible
- Washing chamber and internal door is made of AISI 316L stainless steel
- External panels are made of AISI 304 stainless steel
- Two independent washing levels through telescopic guides
- Contains water softener with salt sensor
- Cold water and demineralized water connections
- Contains no trolleys
- Programs: 9 default, 6 free
- External dimensions: L x D x H: 600 x 605 x 850mm
- Internal working dimensions LxDxH: 525 x 490 x 570mm
Specifica
Specifica
| Product Type | Glassware Washer with Steam Condenser |
| Modello | GW1160SC |
| Requisiti elettrici | 3/N/PE 7kW max |
| Tensione | 400 V |
| Frequenza | 50 Hz |