Learn More
Labconco™ Glassware Washer Rack
Codice prodotto. 17122352 Sfoglia Tutto Labconco Corporation Prodotti
Click to view available options
Descrizione:
Lower Small Spindle Rack
Lower Spindle Rack
Lower Standard Rack
Upper Small Spindle Rack, FlaskScrubber and FlaskScrubber Vantage
Upper Small Spindle Rack, SteamScrubber
Upper Spindle Rack, FlaskScrubber and FlaskScrubber Vantage
Upper Spindle Rack, SteamScrubber
Upper Standard Rack, FlaskScrubber and FlaskScrubber Vantage
Upper Standard Rack, SteamScrubber
Dimensione della confezione:
Pezzo
Codice prodotto. 17122352
Marca: Labconco™ 4668500
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Racks for use with Labconco Glassware Washers with CleanWorks OS
  • Racks and accessories for use with Labconco FlaskScrubber, SteamScrubber, FlaskScrubber Vantage Glassware Washers.
  • Upper Small Spindle Rack has a high-capacity design with 60 spindles, 60 spindle plugs, 12 spindle arm plugs, spindle tool & wash arm for cleaning of small narrow-neck labware in Labconco Glassware Washers. 304 stainless steel. Catalog number for FlaskScrubber & FlaskScrubber Vantage includes upper slide rails for installation.
  • Lower Small Spindle Rack has a high-capacity design with 60 spindles, 60 spindle plugs, 12 spindle arm plugs, spindle tool & wash arm for cleaning of small narrow-neck labware. May be used in place of any Lower Rack in Labconco Glassware Washers. 304 stainless steel.
  • Upper Spindle Rack is designed for narrow-neck labware in Labconco Glassware Washers. Includes 36 spindles, 36 adjust height clips, 18 glassware holders, 10 small spindles & clips, 36 spindle plugs, 12 spindle arm plugs. 304 stainless steel. Catalog number for FlaskScrubber & FlaskScrubber Vantage includes upper slide rails for installation.
  • Upper Standard Rack is open to accommodate accessory inserts to suit your labware needs (sold separately). Type 304 stainless steel. Includes wash arm. May be used in any Labconco Glassware Washer. Catalog number for FlaskScrubber & FlaskScrubber Vantage includes upper slide rails for installation.
  • Lower Spindle Rack is designed for narrow-neck labware. Includes 36 spindles, 36 adjust height clips, 18 glassware holders, 10 small spindles & clips, 36 spindle plugs, 12 spindle arm plugs. Type 304 stainless steel. One is included with each FlaskScrubber or FlaskScrubber Vantage. May be used in place of Lower Standard Rack in SteamScrubber washers.
  • Lower Standard Rack is open to accommodate accessory inserts to suit your labware needs (sold separately). Type 304 stainless steel. Included with every SteamScrubber. May be used in place of Lower Spindle Rack in any FlaskScrubber or FlaskScrubber Vantage Washer.
Specifica
Descrizione Upper Spindle Rack, FlaskScrubber and FlaskScrubber Vantage
Materiale Stainless Steel
Product Type Rack
Tipo di rack Upper Spindle Rack, FlaskScrubber and FlaskScrubber Vantage
Da utilizzare con (apparecchiatura) Labconco FlaskScrubber or FlaskScrubber Vantage
Profondità (sistema imperiale) 21.6 in.
Profondità (sistema metrico) 54.9 cm
Altezza (sistema imperiale) 6.9 in.
Altezza (sistema metrico) 17.5 cm
Lunghezza (sistema imperiale) 21.6 in.
Lunghezza (sistema metrico) 54.9 cm
Larghezza (sistema imperiale) 20.8 in.
Larghezza (sistema metrico) 52.8 cm
Peso (sistema imperiale) di spedizione 23 lb.
Peso (sistema metrico) di spedizione 10.4 kg
Quantità 1/Ea.