Learn More
Thermo Scientific Chemicals Glassy Carbon Boat;Vol (ml), 3;Length (mm), 53;Width (mm), 16;Ht (mm),10
Codice prodotto. 15416295
Click to view available options
Dimensioni (L x P x A):
29 x 16 x 10 mm
53 x 16 x 10 mm
76 x 33 x 25 mm
103 x 16 x 10 mm
Capacità (sistema metrico):
2 mL
3 mL
10 mL
40 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 15416295
Marca: Thermo Scientific Chemicals 039027.KT
Richiesta di formato massivo o personalizzato
Offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Resistance to all wet decomposition agents and abrasive wear
  • No memory effects or contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Resistance to thermal shock allows rapid heating and cooling times
  • Good electrical conductivity
  • Used in vessels for ultra-high purity materials technology such as semiconductor connections and crystal­ growing such as doped halogenide crystals
TRUSTED_SUSTAINABILITY
Specifica
Tipo Glassy Carbon Boat
Capacità (sistema metrico) 3 mL
Dimensioni (L x P x A) 53 x 16 x 10 mm
Altezza (sistema metrico) 10 mm
Lunghezza (sistema metrico) 53 mm
Larghezza (sistema metrico) 16 mm