Learn More
SICCO Gloves
Click to view available options
Size:
7
8
9
Dimensione della confezione:
1 paio
Descrizione
Wearable on both sides, suitable for openings of 190 mm, resistant to tear, perforation and abrasion, high flexibility, low voltage value, minimal static charge. Scope of delivery: 1 pair
Specifica
Specifica
| Colore | Yellow |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Product Type | Gloves |
| Size | 7 |
| Lunghezza (sistema metrico) | 750 mm |