Learn More
Glutathione Peroxidase 1/GPX1 Antibody (3G1), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-26546-50ul
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Glutathione Peroxidase 1/GPX1 Monoclonal antibody specifically detects Glutathione Peroxidase 1/GPX1 in Human samples. It is validated for ELISA
Specifica
| Glutathione Peroxidase 1/GPX1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 3G1 | |
| ELISA | |
| Cellular glutathione peroxidase, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GSHPx-1, GSHPX1, MGC14399, MGC88245 | |
| A synthesized peptide derived from Human Glutathione Peroxidase 1/GPX1 [UniProt P07203] | |
| 50 μL | |
| Apoptosis, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 2876 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |