missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ GlycanTune™ B+ Total Feed

Codice prodotto. 30283911
Click to view available options
Quantità:
1 L
10 L
100 L
Dimensione della confezione:
100L
10L
1L
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30283911

Marca: Gibco™ A2972004

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

GlycanTune™ B+ Total Feed is an AGT™, or Advanced Granulation Technology™, feeding supplement specifically designed for a fed-batch culture of multiple cell lines.

GlycanTune™ B+ Total Feed is an AGT™, or Advanced Granulation Technology™, feeding supplement specifically designed for a fed-batch culture of multiple cell lines. When used, this dry-format, single-part total feeding supplement can shift glycan profiles from heavily G0F to mostly G1F and G2F glycans. The product is an animal origin–free, chemically-defined formulation that contains no proteins, hydrolysates, or components of unknown composition. GlycanTune B+ Total Feed provides an easy-to-reconstitute, pH-neutral solution that can be concentrated from the suggested 59.2 g/L (1X) up to 177.6 g/L (3X). As a result, key nutrients can be delivered in smaller volumes.

Which GlycanTune Total Feed to choose?

Consider GlycanTune B+ Total Feed when:

  • You have used Gibco™ EfficientFeed™ B+ (Plus) AGT Supplement in the past and wish to maintain the benefits of simpler reconstitution and super-concentration in delivering key components alongside improved glycan profile
  • You are currently using another feed and wish to shift glycan profile more toward G1 and G2
  • You are using a cell line or base medium where CD CHO Medium may be considered an option

Consider GlycanTune A+ Total Feed (A2971904) when:

  • You have used Gibco EfficientFeed A+ (Plus) AGT Supplement in the past and wish to maintain the benefits of simpler reconstitution and super-concentration in delivering key components alongside improved glycan profile
  • You are currently using another feed and wish to shift glycan profile more toward G1 and G2
  • You are using a cell line or base medium where CD OptiCHO™ Medium may be considered an option

Consider GlycanTune C+ Total Feed (A2972104) when:

  • You have used Gibco EfficientFeed C+ (Plus) AGT Supplement in the past and wish to maintain the benefits of simpler reconstitution and super-concentration in delivering key components alongside improved glycan profile
  • You are currently using another feed and wish to shift glycan profile more toward G1 and G2
  • You are using a cell line or base medium where Dynamis™ Medium may be considered an option

Dialing in a glycan profile

By following the protocol recommended to test with GlycanTune Total Feed, it is possible to shift the glycan protein expression profiles from primarily G0 to primarily G1 and G2.

Increasing titers

As a complete feed, GlycanTune total feeds have been shown to provide multiple-fold increases in titers at 1X concentration versus batch culture; as such, they are an excellent starting point for process development when a shift in glycan profile is desired. Flexibility to further increase titers is enhanced with the ability to super-concentrate the product by reconstituting with less water to deliver up to 177.6 g/L of fed nutrients (see figure).

Maximizing bioreactor utilization

Have you ever wished you could have more working volume at the start of a bioreactor run to get more protein produced, but needed too much feeding volume space to accomplish your optimal goal? Have you ever thought that room for a little more feeding might get you to a new level of titer from a process and maybe even reduce the number of bioreactor runs that might be needed per year? In either of these cases, it is important to have the flexibility to super-concentrate your feeding supplements so you get less product dilution, more space for an additional feeding, and/or more working volume to start your run (see figure). In fact, you may gain about 20% more bioreactor utilization by exploring the unique ability to super-concentrate this GlycanTune B+ product.

Simplified handling to reduce risk in scale-up

The ability to scale up using AGT dry format provides easier handling and storage in manufacturing and other bioprocess settings. Fewer parts and reconstitution steps mean fewer opportunities for operator error. Further, the ability to store this feed, even when super-concentrated, for the length of a bioreactor run at room temperature in the dark may allow less chance of contamination by reducing the number of connections to the bioreactor to one or two—depending on whether GlycanTune Total Feed can be used alone or after a change from your current feed.

TRUSTED_SUSTAINABILITY

Specifica

Content And Storage AGT™ dry format. Store in refrigerator (2–8°C). Protect from light and moisture.
Product Type Microbiology Media
Linea di prodotti GlycanTune
Quantità 1 L
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.